Anti-S-Tag Epitope Tag Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB600-511
102134-168EA 564.74 USD
102134-168
Anti-S-Tag Epitope Tag Rabbit Polyclonal Antibody
Antibodies
The S-Tag Epitope Tag Antibody from Novus Biologicals is a rabbit polyclonal antibody to S-Tag Epitope Tag. This antibody reacts with non-species specific. The S-Tag Epitope Tag Antibody has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunoprecipitation.

Type: Primary
Antigen: S-Tag Epitope Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Non-species specific
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR