Anti-S-Tag Epitope Tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

NB600-512
102134-172EA 564.62 USD
102134-172
Anti-S-Tag Epitope Tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Rabbit Polyclonal S-Tag Epitope Tag Antibody [HRP]. Tested Applications: Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (-). Tested Reactivity: Non-species specific.

Type: Primary
Antigen: S-Tag Epitope Tag
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR