Anti-ST8SIA2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-69280
102198-800EA 564.74 USD
102198-800
Anti-ST8SIA2 Rabbit Polyclonal Antibody
Antibodies
The ST8 alpha-2,8-Sialyltransferase 8B / ST8SIA2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to ST8 alpha-2,8-Sialyltransferase 8B / ST8SIA2. This antibody reacts with human. The ST8 alpha-2,8-Sialyltransferase 8B / ST8SIA2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: ST8SIA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR