Anti-ST6GAL2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-74154
102205-050EA 564.74 USD
102205-050
Anti-ST6GAL2 Rabbit Polyclonal Antibody
Antibodies
The ST6 Gal Sialyltransferase 2 / ST6GAL2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to ST6 Gal Sialyltransferase 2 / ST6GAL2. This antibody reacts with mouse. The ST6 Gal Sialyltransferase 2 / ST6GAL2 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: ST6GAL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR