Anti-SSH1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-60673
102111-096EA 635.51 USD
102111-096
Anti-SSH1 Rabbit Polyclonal Antibody
Antibodies
The SSH1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SSH1. This antibody reacts with human. The SSH1 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: SSH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR