Anti-SRPX2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-50176
102180-148EA 329 USD
102180-148
Anti-SRPX2 Rabbit Polyclonal Antibody
Antibodies
The SRPX2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SRPX2. This antibody reacts with human, monkey. The SRPX2 Antibody has been validated for the following applications: Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 10 ?g/ml

Type: Primary
Antigen: SRPX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR