Anti-SRPK1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-30108
102162-962EA 635.51 USD
102162-962
Anti-SRPK1 Rabbit Polyclonal Antibody
Antibodies
The SRPK1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SRPK1. This antibody reacts with human, mouse. The SRPK1 Antibody has been validated for the following applications: Immunohistochemistry.

Type: Primary
Antigen: SRPK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR