Anti-SRP19 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-57415
102189-722EA 564.74 USD
102189-722
Anti-SRP19 Rabbit Polyclonal Antibody
Antibodies
The SRP19 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SRP19. This antibody reacts with human. The SRP19 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry.

Type: Primary
Antigen: SRP19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR