Anti-SRP1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-33508
102167-106EA 635.51 USD
102167-106
Anti-SRP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal SRP1 Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence. Tested Reactivity: Human.

Type: Primary
Antigen: SRP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR