Anti-SRD5A2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-69492
102199-054EA 564.74 USD
102199-054
Anti-SRD5A2 Rabbit Polyclonal Antibody
Antibodies
The SRD5A2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SRD5A2. This antibody reacts with human, monkey. The SRD5A2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry.

Type: Primary
Antigen: SRD5A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR