Anti-SRD5A2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-46510
102176-300EA 564.74 USD
102176-300
Anti-SRD5A2 Rabbit Polyclonal Antibody
Antibodies
The SRD5A2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SRD5A2. This antibody reacts with human, mouse. The SRD5A2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SRD5A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR