Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SR-BI Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Novus Biologicals
NB400-104H
The SR-BI Antibody [HRP] from Novus Biologicals is a rabbit polyclonal antibody to SR-BI. This antibody reacts with human, mouse, rat, chinese hamster, hamster, mustelid, primate, rabbit, golden syrian hamster. The SR-BI Antibody [HRP] has been validated for the following applications: Western Blot, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Type: Primary
Antigen: SR-BI
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Chinese Hamster, Hamster, Mustelid, Primate, Rabbit, Golden Syrian Hamster




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μl NB400-104H 102131-322 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol SR-BI
Antigen Name Scavenger Receptor Class B, Member 1
Reactivity Primate, Golden Syrian Hamster, Rat, Hamster, Chinese Hamster, Human, Mouse, Mustelid, Rabbit
Conjugation HRP (Horseradish Peroxidase)
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
Antigen Synonyms scavenger receptor class B type III, SR-B1, CLA1 CD36 antigen-like 1, CD36 antigen (collagen type I receptor, CD36 and LIMPII analogous 1, thrombospondin receptor-like 1, Collagen type I receptor, SCARB1, CD36 antigen, thrombospondin receptor)-like 1, scavenger receptor class B, SRBI, MGC138242, CD36L1scavenger receptor class B type 1, CLA-1 HDLQTL6, SRB1 scavenger receptor class B member 1, member 1
Gene ID 949
Immunogen A C-terminal peptide containing residues from mouse SR-BI (within residues 450-509). [UniProt# Q61009]
Purification Immunogen affinity purified
Storage Buffer PBS
Storage Temperature Store at 4C in the dark.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty