Anti-SRBD1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-57557
102189-996EA 564.74 USD
102189-996
Anti-SRBD1 Rabbit Polyclonal Antibody
Antibodies
The SRBD1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SRBD1. This antibody reacts with human. The SRBD1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry.

Type: Primary
Antigen: SRBD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR