Anti-SRA1 Rabbit Polyclonal Antibody

NB100-2270
102106-962EA 536.92 USD
102106-962
Anti-SRA1 Rabbit Polyclonal Antibody
The SRA1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SRA1. This antibody reacts with human. The SRA1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SRA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR