Anti-SR1 Rabbit Polyclonal Antibody

NBP1-32340
102165-804EA 493.85 USD
102165-804
Anti-SR1 Rabbit Polyclonal Antibody
The SR1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SR1. This antibody reacts with human, mouse. The SR1 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: SR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR