Anti-Spt6 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-2582
102107-828EA 635.51 USD
102107-828
Anti-Spt6 Rabbit Polyclonal Antibody
Antibodies
The Spt6 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Spt6. This antibody reacts with human, mouse, chicken. The Spt6 Antibody has been validated for the following applications: Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100 - 1:500

Type: Primary
Antigen: Spt6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR