Anti-SPRY4 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-2032
102106-536EA 777.05 USD
102106-536
Anti-SPRY4 Rabbit Polyclonal Antibody
Antibodies
The SPRY4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SPRY4. This antibody reacts with human. The SPRY4 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SPRY4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR