Anti-SPRY2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-04100
102234-914EA 635.51 USD
102234-914
Anti-SPRY2 Rabbit Polyclonal Antibody
Antibodies
The SPRY2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SPRY2. This antibody reacts with human. The SPRY2 Antibody has been validated for the following applications: Immunoprecipitation.

Type: Primary
Antigen: SPRY2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR