Anti-SPNS2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-54345
102183-960EA 564.74 USD
102183-960
Anti-SPNS2 Rabbit Polyclonal Antibody
Antibodies
The SPNS2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SPNS2. This antibody reacts with human. The SPNS2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 5 ?g/ml

Type: Primary
Antigen: SPNS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR