Anti-Sphingosine Kinase 2/SPHK2 Rabbit Polyclonal Antibody

NBP1-50777
102180-584EA 466.15 USD
102180-584
Anti-Sphingosine Kinase 2/SPHK2 Rabbit Polyclonal Antibody
The Sphingosine Kinase 2 / SPHK2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Sphingosine Kinase 2 / SPHK2. This antibody reacts with human, mouse, rat. The Sphingosine Kinase 2 / SPHK2 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Sphingosine Kinase 2/SPHK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR