Anti-Sphingosine Kinase 1/SPHK1 Rabbit Polyclonal Antibody (DyLight 488)

Supplier: Novus Biologicals
NBP1-22974G
102160-784EA 706.28 USD
102160-784
Anti-Sphingosine Kinase 1/SPHK1 Rabbit Polyclonal Antibody (DyLight 488)
Antibodies
The Sphingosine Kinase 1 / SPHK1 Antibody [DyLight 488] from Novus Biologicals is a rabbit polyclonal antibody to Sphingosine Kinase 1 / SPHK1. This antibody reacts with human. The Sphingosine Kinase 1 / SPHK1 Antibody [DyLight 488] has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Sphingosine Kinase 1/SPHK1
Clonality: Polyclonal
Clone:
Conjugation: DyLight 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR