Anti-SPG8 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
45530002
102104-138EA 329 USD
102104-138
Anti-SPG8 Rabbit Polyclonal Antibody
Rabbit Polyclonal SPG8 Antibody. Tested Applications: Western Blot, ELISA. Tested Reactivity: Human.

Type: Primary Antibodies
Antigen: SPG8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR