Anti-SPG11 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-76990
102210-164EA 564.74 USD
102210-164
Anti-SPG11 Rabbit Polyclonal Antibody
Antibodies
The SPG11 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SPG11. This antibody reacts with human, mouse, rat. The SPG11 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SPG11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR