Anti-Spastin Rabbit Polyclonal Antibody

NB100-62922
102112-140EA 423.08 USD
102112-140
Anti-Spastin Rabbit Polyclonal Antibody
The Spastin Antibody from Novus Biologicals is a rabbit polyclonal antibody to Spastin. This antibody reacts with human. The Spastin Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: Spastin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR