Anti-TFF2 Rabbit Polyclonal Antibody

NBP1-45886
102175-306EA 466.15 USD
102175-306
Anti-TFF2 Rabbit Polyclonal Antibody
The TFF2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TFF2. This antibody reacts with human. The TFF2 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TFF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR