Anti-SPANXC Rabbit Polyclonal Antibody

NBP1-77925
102212-230EA 564.62 USD
102212-230
Anti-SPANXC Rabbit Polyclonal Antibody
The SPANXC Antibody from Novus Biologicals is a rabbit polyclonal antibody to SPANXC. This antibody reacts with human. The SPANXC Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SPANXC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR