Anti-SPANXC Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-03285
102155-796EA 706.28 USD
102155-796
Anti-SPANXC Rabbit Polyclonal Antibody
Antibodies
The SPANXC Antibody from Novus Biologicals is a rabbit polyclonal antibody to SPANXC. This antibody reacts with human, primate. The SPANXC Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 5 ?g/ml

Type: Primary
Antigen: SPANXC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR