Anti-SPAG9 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-31153
102164-216EA 635.51 USD
102164-216
Anti-SPAG9 Rabbit Polyclonal Antibody
Antibodies
The SPAG9 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SPAG9. This antibody reacts with human. The SPAG9 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SPAG9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR