Anti-SPAG11B Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-91448
102217-434EA 564.74 USD
102217-434
Anti-SPAG11B Rabbit Polyclonal Antibody
Antibodies
The SPAG11B Antibody from Novus Biologicals is a rabbit polyclonal antibody to SPAG11B. This antibody reacts with human. The SPAG11B Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SPAG11B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR