Anti-SP140L Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-80140
102215-928EA 564.74 USD
102215-928
Anti-SP140L Rabbit Polyclonal Antibody
Antibodies
The SP140L Antibody from Novus Biologicals is a rabbit polyclonal antibody to SP140L. This antibody reacts with human. The SP140L Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SP140L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR