Anti-SOX7 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-80374
102216-396EA 564.74 USD
102216-396
Anti-SOX7 Rabbit Polyclonal Antibody
Antibodies
The SOX7 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SOX7. This antibody reacts with human. The SOX7 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SOX7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR