Anti-SOX6 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-59987
102124-008EA 279 USD
102124-008
Anti-SOX6 Rabbit Polyclonal Antibody
Antibodies
The SOX6 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SOX6. This antibody reacts with human, mouse, rat, primate. The SOX6 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:300

Type: Primary
Antigen: SOX6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR