Anti-SOX17 Rabbit Polyclonal Antibody

NBP1-80020
102215-688EA 466.15 USD
102215-688
Anti-SOX17 Rabbit Polyclonal Antibody
The SOX17 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SOX17. This antibody reacts with human. The SOX17 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SOX17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR