Anti-SOS1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-05954
102156-950EA 635.51 USD
102156-950
Anti-SOS1 Rabbit Polyclonal Antibody
Antibodies
The SOS1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SOS1. This antibody reacts with human. The SOS1 Antibody has been validated for the following applications: Immunoprecipitation.

Type: Primary
Antigen: SOS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR