Anti-SorCS3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-30615
102163-680EA 564.74 USD
102163-680
Anti-SorCS3 Rabbit Polyclonal Antibody
Antibodies
The SorCS3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SorCS3. This antibody reacts with human. The SorCS3 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Recommended Dilutions: Immunohistochemistry (Parafin): 10-50 ?g/ml

Type: Primary
Antigen: SorCS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR