Anti-Shh Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-69270
102198-784EA 635.51 USD
102198-784
Anti-Shh Rabbit Polyclonal Antibody
Antibodies
The Sonic Hedgehog / Shh Antibody from Novus Biologicals is a rabbit polyclonal antibody to Sonic Hedgehog / Shh. This antibody reacts with human, chicken. The Sonic Hedgehog / Shh Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:1000

Type: Primary
Antigen: Shh
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR