Anti-SSTR5 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NLS4419
102253-364EA 564.74 USD
102253-364
Anti-SSTR5 Rabbit Polyclonal Antibody
Antibodies
The Somatostatin R5 / SSTR5 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Somatostatin R5 / SSTR5. This antibody reacts with human. The Somatostatin R5 / SSTR5 Antibody has been validated for the following applications: Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SSTR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR