Anti-SSTR4 Rabbit Polyclonal Antibody

NLS4148
102253-338EA 493.85 USD
102253-338
Anti-SSTR4 Rabbit Polyclonal Antibody
The Somatostatin R4 / SSTR4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Somatostatin R4 / SSTR4. This antibody reacts with human. The Somatostatin R4 / SSTR4 Antibody has been validated for the following applications: Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 3 ?g/ml to 17 ?g/ml

Type: Primary
Antigen: SSTR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR