Anti-SSTR4 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-74539
102116-466EA 329 USD
102116-466
Anti-SSTR4 Rabbit Polyclonal Antibody
Antibodies
The Somatostatin R4 / SSTR4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Somatostatin R4 / SSTR4. This antibody reacts with human, mouse. The Somatostatin R4 / SSTR4 Antibody has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:2500

Type: Primary
Antigen: SSTR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR