Anti-SSTR3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-74538
102116-464EA 564.74 USD
102116-464
Anti-SSTR3 Rabbit Polyclonal Antibody
Antibodies
The Somatostatin R3 / SSTR3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Somatostatin R3 / SSTR3. This antibody reacts with human. The Somatostatin R3 / SSTR3 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:2500

Type: Primary
Antigen: SSTR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR