Anti-SST Rabbit Polyclonal Antibody

NB110-61682
102124-542EA 395.38 USD
102124-542
Anti-SST Rabbit Polyclonal Antibody
Rabbit Polyclonal Somatostatin Antibody. Tested Applications: ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Radioimmunoassay. Tested Reactivity: All Species.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:1000

Type: Primary
Antigen: SST
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR