Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Solute carrier family 22 member 18 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-59747
The Solute carrier family 22 member 18 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Solute carrier family 22 member 18. This antibody reacts with human. The Solute carrier family 22 member 18 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Solute carrier family 22 member 18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μl NBP1-59747 102193-180 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol Solute carrier Family 22 member 18
Antigen Name Solute carrier family 22 member 18
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
Antigen Synonyms Efflux transporter-like protein, IMPT1DKFZp667A184, HET, Tumor-suppressing subchromosomal transferable fragment candidate gene 5 protein, ORCTL-2, organic cation transporter-like 2, member 1-like, Tumor-suppressing STF cDNA 5 protein, BWSCR1Asolute carrier family 22 (organic cation transporter), Organic cation transporter-like protein 2, imprinted multi-membrane spanning polyspecific transporter-related protein 1, ITMp45-BWR1A, solute carrier family 22, solute carrier family 22 member 18, member 18, Imprinted multi-membrane-spanning polyspecific transporter-related protein 1, BWR1AORCTL2, Solute carrier family 22 member 1-like, p45 Beckwith-Wiedemann region 1A, SLC22A1Lcandidate A, TSSC5p45-Beckwith-Wiedemann region 1 A
Gene ID 5002
Immunogen Synthetic peptides corresponding to SLC22A18(solute carrier family 22, member 18) The peptide sequence was selected from the N terminal of SLC22A18. Peptide sequence AASSPALPGVYLLFASRLPGALMHTLPAAQMVITDLSAPEERPAALGRLG.
Purification Immunogen affinity purified
Storage Buffer PBS and 2% Sucrose
Storage Temperature Store at -20C. Avoid freeze-thaw cycles.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty