Anti-Soluble Liver/Pancreas Antigen Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-57253
102189-402EA 564.74 USD
102189-402
Anti-Soluble Liver/Pancreas Antigen Rabbit Polyclonal Antibody
Antibodies
The Soluble Liver / Pancreas Antigen Antibody from Novus Biologicals is a rabbit polyclonal antibody to Soluble Liver / Pancreas Antigen. This antibody reacts with human. The Soluble Liver / Pancreas Antigen Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Soluble Liver/Pancreas Antigen
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR