Anti-SOHLH1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-80284
102216-216EA 564.74 USD
102216-216
Anti-SOHLH1 Rabbit Polyclonal Antibody
Antibodies
The SOHLH1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SOHLH1. This antibody reacts with mouse. The SOHLH1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SOHLH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR