Anti-SLC5A5 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-59851
102193-386EA 564.74 USD
102193-386
Anti-SLC5A5 Rabbit Polyclonal Antibody
Antibodies
The SLC5A5 / Sodium Iodide Symporter Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC5A5 / Sodium Iodide Symporter. This antibody reacts with human. The SLC5A5 / Sodium Iodide Symporter Antibody has been validated for the following applications: Western Blot, Immunohistochemistry.

Type: Primary
Antigen: SLC5A5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR