Anti-SOCS-7 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-58902
102191-526EA 564.74 USD
102191-526
Anti-SOCS-7 Rabbit Polyclonal Antibody
Antibodies
The SOCS-7 / Nck / NAP4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SOCS-7 / Nck / NAP4. This antibody reacts with human. The SOCS-7 / Nck / NAP4 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SOCS-7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR