Anti-SOCS-2 Rabbit Polyclonal Antibody

NB600-1338
102133-222EA 395.38 USD
102133-222
Anti-SOCS-2 Rabbit Polyclonal Antibody
The SOCS-2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SOCS-2. This antibody reacts with human, mouse, rat. The SOCS-2 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SOCS-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR