Anti-SOCS-1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-56637
102110-192EA 491.08 USD
102110-192
Anti-SOCS-1 Rabbit Polyclonal Antibody
Antibodies
The SOCS-1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SOCS-1. This antibody reacts with human, mouse. The SOCS-1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SOCS-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR