Anti-SNURPORTIN1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-57132
102189-166EA 564.74 USD
102189-166
Anti-SNURPORTIN1 Rabbit Polyclonal Antibody
Antibodies
The SNURPORTIN1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SNURPORTIN1. This antibody reacts with human. The SNURPORTIN1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SNURPORTIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR