Anti-SNRPF Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-57463
102189-818EA 564.74 USD
102189-818
Anti-SNRPF Rabbit Polyclonal Antibody
Antibodies
The SNRPF Antibody from Novus Biologicals is a rabbit polyclonal antibody to SNRPF. This antibody reacts with human. The SNRPF Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SNRPF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR